PRIH93ED7B8C Princeton BioMolecular Research, Inc.

(4E)-4-benzylidene-2-phenyl-1,3-oxazol-5(4H)-one

  • MDL number MFCD00098978
  • Cas Number : 842-74-0
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart