PRIH93ED7B90 Princeton BioMolecular Research, Inc.

5-(2,4-Dimethoxy-phenyl)-2H-pyrazole-3-carboxylic acid

  • MDL number MFCD03030178
  • Cas Number : 879765-63-6
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart