PRIH93ED7B93 Princeton BioMolecular Research, Inc.

1',3',5'-Trimethyl-1H,1'H-[3,4']bipyrazolyl-5-carboxylic acid

  • MDL number MFCD07186517
  • Cas Number : 890624-34-7
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart