PRIH93ED7B97 Princeton BioMolecular Research, Inc.

N-(2,5-Dimethyl-phenyl)-benzenesulfonamide

  • MDL number MFCD01212620
  • Cas Number : 88681-01-0
  • Molecular Weight : 261.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart