PRIH93ED7BA1 Princeton BioMolecular Research, Inc.

7-(bromomethyl)-4,7-dimethylspiro[bicyclo[2.2.1]heptane-2,3,di2'-[1,3]dioxolan]

  • MDL number MFCD09910520
  • Molecular Weight : 333.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart