PRIH93ED7BA2 Princeton BioMolecular Research, Inc.

1,7-dibromo-4-(dibromomethyl)-3,3-dimethylbicyclo[2.2.1]heptan-2-one

  • MDL number MFCD00702836
  • Molecular Weight : 467.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart