PRIH93ED7BA5 Princeton BioMolecular Research, Inc.

11-(bromomethyl)-1,11-dimethyl-1,2,3,4-tetrahydro-1,4-methanophenazine

  • MDL number MFCD04066533
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart