PRIH93ED7BA8 Princeton BioMolecular Research, Inc.

2-chloro-5-(morpholin-4-ylsulfonyl)benzoic acid

  • MDL number MFCD00492872
  • Cas Number : 109029-96-1
  • Molecular Weight : 305.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart