PRIH93ED7BBD Princeton BioMolecular Research, Inc.

7-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]quinolin-1-amine

  • MDL number MFCD06496308
  • Molecular Weight : 199.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart