PRIH93ED7BD3 Princeton BioMolecular Research, Inc.

2-ethyl-1'H-1,2'-bibenzimidazole

  • MDL number MFCD11108425
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart