PRIH93ED7BF3 Princeton BioMolecular Research, Inc.

4-bromo-3,5-dimethyl-2-furoic acid

  • MDL number MFCD11557222
  • Cas Number : 1092286-18-4
  • Molecular Weight : 219.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart