PRIH93ED7BF7 Princeton BioMolecular Research, Inc.

2-bromo-1H-benzimidazole-5-carboxylic acid

  • MDL number MFCD09837250
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart