PRIH93ED7BFB Princeton BioMolecular Research, Inc.

2-(1-methylhydrazino)propanoic acid

  • MDL number MFCD11557290
  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart