PRIH93ED7BFE Princeton BioMolecular Research, Inc.

2-(3,5-Dimethyl-pyrazol-1-yl)-benzothiazole

  • MDL number MFCD00099325
  • Cas Number : 23615-60-3
  • Molecular Weight : 229.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart