PRIH93ED7C0E Princeton BioMolecular Research, Inc.

6-ethyl-6H-indolo[2,3-b]quinoxaline

  • MDL number MFCD00812200
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart