PRIH93ED7C1D Princeton BioMolecular Research, Inc.

[(1-isopropyl-1H-benzimidazol-2-yl)thio]acetic acid

  • MDL number MFCD00106142
  • Cas Number : 263147-78-0
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart