PRIH93ED7C47 Princeton BioMolecular Research, Inc.

5-Methoxymethyl-7-(4-methyl-piperidin-1-ylmethyl)-quinolin-8-ol

  • MDL number MFCD00468375
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart