PRIH93ED7C5F Princeton BioMolecular Research, Inc.

Formic acid formyloxy-(2-nitro-phenyl)-methyl ester

  • MDL number MFCD12972307
  • Molecular Weight : 225.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart