PRIH93ED7C81 Princeton BioMolecular Research, Inc.

2-{4-[3-Oxo-3-(4-propoxy-phenyl)-propylamino]-benzyl}-tetrahydro-pyrrolo[1,2-c]imidazole-1,3-dione

  • MDL number MFCD12972546
  • Molecular Weight : 435.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart