PRIH93ED7C84 Princeton BioMolecular Research, Inc.

4-Benzo[1,3]dioxol-5-yl-[1,2,4]triazolidine-3,5-dione

  • MDL number MFCD12033726
  • Molecular Weight : 221.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart