PRIH93ED7C87 Princeton BioMolecular Research, Inc.

3-(2-Adamantan-1-yl-ethyl)-2-(4-chloro-phenylimino)-4-oxo-[1,3]thiazinane-6-carboxylic acid

  • MDL number MFCD13704595
  • Molecular Weight : 447

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart