PRIH93ED7C8B Princeton BioMolecular Research, Inc.

pyrimidine, 2-[[[2,3-dimethyl-6-[(2-pyrimidinylthio)methyl]phenyl]methyl]thio]-

  • MDL number MFCD13704635
  • Molecular Weight : 354.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart