PRIH93ED7C8D Princeton BioMolecular Research, Inc.

1,3,5-Trichloro-2,4,6-trinitro-benzene

  • MDL number MFCD00194304
  • Cas Number : 2631-68-7
  • Molecular Weight : 316.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart