PRIH93ED7C92 Princeton BioMolecular Research, Inc.

2H-indol-2-one, 1,3-dihydro-5,6-dimethoxy-3-[(2,3,4-trimethoxyphenyl)methylene]-, (3Z)-

  • MDL number MFCD12034284
  • Molecular Weight : 371.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart