PRIH93ED7CA4 Princeton BioMolecular Research, Inc.

4,5,6-Trimethoxy-2-(3,4,5-trimethoxy-phenyl)-indan-1,3-dione

  • MDL number MFCD12033843
  • Molecular Weight : 402.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart