PRIH93ED7CA7 Princeton BioMolecular Research, Inc.

2-Oxo-3-(5-p-tolyl-[1,3,4]oxadiazol-2-ylmethyl)-2,3-dihydro-benzooxazole-5-carboxylic acid methyl ester

  • MDL number MFCD12972478
  • Molecular Weight : 365.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart