PRIH93ED7CB2 Princeton BioMolecular Research, Inc.

2-(2,3-Dimethoxy-benzylidene)-malononitrile

  • MDL number MFCD00449704
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart