PRIH93ED7CC1 Princeton BioMolecular Research, Inc.

4(1H)-quinazolinone, 3-(1,3-benzodioxol-5-yl)-2,3-dihydro-2-(5-hydroxy-3-methyl-1-phenyl-1H-pyrazol-4-yl)-

  • MDL number MFCD13704948
  • Molecular Weight : 440.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart