PRIH93ED7CC7 Princeton BioMolecular Research, Inc.

3-(2-Hydroxy-3-methoxy-benzylidene)-5,6-dimethoxy-1,3-dihydro-indol-2-one

  • MDL number MFCD03852839
  • Molecular Weight : 327.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart