PRIH93ED7CD5 Princeton BioMolecular Research, Inc.

carbamic acid, [4-[[4-[[(2-chloroethoxy)carbonyl]amino]phenyl]dithio]phenyl]-, 2-chloroethyl ester

  • MDL number MFCD13705045
  • Molecular Weight : 461.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart