PRIH93ED7CD7 Princeton BioMolecular Research, Inc.

3-(3-Furan-2-ylmethyl-2-oxo-6-trifluoromethyl-2,3-dihydro-benzoimidazol-1-ylmethyl)-benzonitrile

  • MDL number MFCD13705055
  • Molecular Weight : 397.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart