PRIH93ED7CDF Princeton BioMolecular Research, Inc.

8-Imidazol-1-yl-1,3,7-trimethyl-3,7-dihydro-purine-2,6-dione

  • MDL number MFCD01580979
  • Molecular Weight : 260.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart