PRIH93ED7CF9 Princeton BioMolecular Research, Inc.

5-{[(2-Chloro-acetylamino)-methyl]-amino}-2-cyano-3-methyl-hexa-2,4-dienoic acid

  • MDL number MFCD12033968
  • Molecular Weight : 271.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart