PRIH93ED7D06 Princeton BioMolecular Research, Inc.

5-Methyl-3-(2-nitro-phenyl)-imidazolidine-2,4-dione

  • MDL number MFCD13705183
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart