PRIH93ED7D11 Princeton BioMolecular Research, Inc.

(4-Diethylcarbamoyl-2-methoxy-6-nitro-phenoxy)-acetic acid ethyl ester

  • MDL number MFCD12033956
  • Molecular Weight : 354.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart