PRIH93ED7D27 Princeton BioMolecular Research, Inc.

1-[2-(2-Methoxy-phenoxy)-ethyl]-2-trifluoromethyl-1H-benzoimidazole

  • MDL number MFCD01141692
  • Molecular Weight : 336.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart