PRIH93ED7D40 Princeton BioMolecular Research, Inc.

2-(6-Nitro-1H-benzoimidazol-2-ylmethylsulfanyl)-benzothiazole

  • MDL number MFCD01785180
  • Molecular Weight : 342.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart