PRIH93ED7D57 Princeton BioMolecular Research, Inc.

1-(2-p-Tolyloxy-ethyl)-2-trifluoromethyl-1H-benzoimidazole

  • MDL number MFCD01141698
  • Molecular Weight : 320.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart