PRIH93ED7D70 Princeton BioMolecular Research, Inc.

2-{1-[2-(2-Methoxy-phenoxy)-ethyl]-1H-benzoimidazol-2-ylsulfanyl}-ethanol

  • MDL number MFCD01163839
  • Molecular Weight : 344.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart