PRIH93ED7D73 Princeton BioMolecular Research, Inc.

2-[2-(4-Methoxy-phenoxy)-ethylsulfanyl]-1H-benzoimidazole

  • MDL number MFCD00808072
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart