PRIH93ED7D86 Princeton BioMolecular Research, Inc.

(1H-Benzoimidazol-2-yl)-hydroxyimino-acetonitrile

  • MDL number MFCD00841115
  • Cas Number : 63823-75-6
  • Molecular Weight : 186.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart