PRIH93ED7D8C Princeton BioMolecular Research, Inc.

4-Chloro-N-phenethyl-benzenesulfonamide

  • MDL number MFCD00708546
  • Molecular Weight : 295.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart