PRIH93ED7D92 Princeton BioMolecular Research, Inc.

N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-methanesulfonamide

  • MDL number MFCD00739478
  • Molecular Weight : 281.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart