PRIH93ED7D9D Princeton BioMolecular Research, Inc.

[2-Nitro-4-(piperidine-1-sulfonyl)-phenoxy]-acetic acid methyl ester

  • MDL number MFCD03819340
  • Molecular Weight : 358.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart