PRIH93ED7DAD Princeton BioMolecular Research, Inc.

3-Methyl-4-oxo-2-(5,6,7-trimethoxy-3-oxo-1,3-dihydro-isobenzofuran-4-ylmethylsulfanyl)-3,4-dihydro-quinazoline-7-carboxylic acid methyl ester

  • MDL number MFCD14856427
  • Molecular Weight : 486.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart