PRIH93ED7DB4 Princeton BioMolecular Research, Inc.

2-(5-Amino-[1,3,4]thiadiazol-2-ylsulfanyl)-1-(1,2,3,4,5,6,7,8-octahydro-phenanthren-9-yl)-ethanone

  • MDL number MFCD15204106
  • Molecular Weight : 359.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart