PRIH93ED7DB9 Princeton BioMolecular Research, Inc.

5-Bromo-2-methylsulfanyl-1H-benzoimidazole

  • MDL number MFCD16872353
  • Molecular Weight : 243.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart