PRIH93ED7DBB Princeton BioMolecular Research, Inc.

2-(1H-Benzoimidazol-2-ylsulfanyl)-acetamide

  • MDL number MFCD00656408
  • Cas Number : 30065-26-0
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart