PRIH93ED7DCB Princeton BioMolecular Research, Inc.

[2-(2-Morpholin-4-ylmethyl-benzoimidazol-1-ylmethyl)-benzoimidazol-1-yl]-acetic acid

  • MDL number MFCD08277443
  • Molecular Weight : 405.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart