PRIH93ED7DCF Princeton BioMolecular Research, Inc.

2-{6-Amino-9-[2-(4-ethoxy-phenoxy)-ethyl]-9H-purin-8-ylsulfanyl}-acetamide

  • MDL number MFCD03272458
  • Molecular Weight : 388.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart